missing translation for 'onlineSavingsMsg'
Learn More

Acetamide, 99%, pure, Thermo Scientific Chemicals

Catalog Number p-4510247
26.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
60-35-5
C2H5NO
59.06
DLFVBJFMPXGRIB-UHFFFAOYSA-N
ethanamide, acetic acid amide, methanecarboxamide, acetimidic acid, amide c2, ethanimidic acid, acetamid, amid kyseliny octove, caswell no. 003h, acetimidic acid van
178
CHEBI:49028
acetamide
CC(=O)N
60-35-5
C2H5NO
59.06
DLFVBJFMPXGRIB-UHFFFAOYSA-N
ethanamide, acetic acid amide, methanecarboxamide, acetimidic acid, amide c2, ethanimidic acid, acetamid, amid kyseliny octove, caswell no. 003h, acetimidic acid van
178
CHEBI:49028
acetamide
CC(=O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 79.0°C to 81.0°C
Color White
Boiling Point 221.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula CH3CONH2
Quantity 100 g
Beilstein 02,II,177
Fieser 01,3
Merck Index 15,44
Solubility Information Solubility in water: 2000g/L (20°C). Other solubilities: 1g/2 mL alcohol- 1g/6 mL pyridine,soluble in chloroform,glycerol and hot benzene,slightly soluble in ether
Formula Weight 59.06
Percent Purity 99%
Grade Pure
Physical Form Crystals
Chemical Name or Material Acetamide
Show More Show Less
compliance-icons
Product Identifier
  • Acetamide
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'