missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetaldehyde, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
2.5 L
Description
Acetaldehyde, Extra Pure, SLR
Specifications
Melting Point | -123°C |
Boiling Point | 21°C |
Packaging | Amber glass bottle |
Quantity | 2.5 L |
Formula Weight | 44.04g/mol |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Acetaldehyde
- Danger
- Flammable liquid Category 1
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
- H224-Extremely flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P235-Store in a well-ventilated place. Keep cool.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'