missing translation for 'onlineSavingsMsg'
Learn More

Acetaldehyde, 99.5%, extra pure, ACROS Organics™

Brand:  Acros Organics 149510010

Code : 29

Additional Details : CAS Number : 75-07-0 Weight : 0.78500kg Transport : UN number : 1089 Chem class : 3 Pack group : I


Packaging Glass bottle
Quantity 1L

 View more versions of this product

Product Code. 10051530

  • £30.65 / 1 L

Estimated Shipment
29-10-2020

Please to purchase this item. Need a web account? Register with us today!



Specifications

Chemical Identifiers

75-07-0
44.04
IKHGUXGNUITLKF-UHFFFAOYSA-N
177
acetaldehyde
C2H4O
MFCD00006991
ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique
CHEBI:15343
CC=O

Specifications

Acetaldehyde
−27°C
Pungent
1.3310 to 1.3330 (20°C, 589nm)
0.02% max.
Colorless
1L
75-07-0
100.0
C2H4O
MFCD00006991
01,3
ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique
IKHGUXGNUITLKF-UHFFFAOYSA-N
acetaldehyde
177
44.04
99.5%
0.7850g/mL
Authentic
Glass bottle
0.785
21.0°C
-123.0°C
Extra Pure
99.4
99.4% min. (GC)
CH3CHO
01, 594
15, 40
Solubility in water: > 500g/L (20°C). Other solubilities: soluble in ethanol and acetone
CC=O
44.04
CHEBI:15343
Volatile Liquid
Extra Pure
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Acetaldehyde

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 1
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Carcinogenicity Category 2

Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P235-Store in a well-ventilated place. Keep cool.

SDS Documents