Simport™ Acetal Co-Polymer Histosette 1 Histology Cassettes with Integral Lid
Overview
Brand: Simport™
Manufacturer Part Number: M490-5
X1500 Histology Cassettes Histosette I With integrBeschriftungsrand, (VE=1500St) gelb
UNSPSC: 41102920
Code: 04
Additional Details:
Additional Details: Weight: 1.00000kg
Product Code. 10334661
Quantity Price
1 £ 157.0 / Pack of 1500
Estimated Shipment
07-03-2017
Add to basket
Description and Specification
Specification
Material Acetal Co-Polymer
Quantity 1500Pack
Color Yellow
Molded from acetal co-polymer, these cassettes keep specimens safely submerged in solvent and are totally resistant to the chemical action of histological solvents. The one-piece integral lid can be opened or closed as often as necessary without danger of specimen loss. Anterior writing area at a 30° angle. Pack quantity 1,500.
Documentation
Technical documents