Learn More
Simport™ Acetal Co-Polymer Histosette 1 Histology Cassettes
Brand:  Simport™ M491-2
UNSPSC : 41102920
Code : 04
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 10664891
  • £143.00 / Pack of 1500
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
Molded from acetal co-polymer, these cassettes keep specimens safely submerged in solvent and are totally resistant to the chemical action of histological solvents. The one-piece integral lid can be opened or closed as often as necessary without danger of specimen loss. Anterior writing area at a 30° angle. Pack quantity 1,500.
Specifications
Specifications
1500Pack
White
Acetal Co-Polymer
Documentation
Technical Documents