missing translation for 'onlineSavingsMsg'
Learn More

Acenaphthenequinone, 95%, Thermo Scientific Chemicals

Catalog Number p-3756749
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
82-86-0
C12H6O2
182.18
MFCD00003805
AFPRJLBZLPBTPZ-UHFFFAOYSA-N
acenaphthenequinone, acenaphthoquinone, 1,2-acenaphthylenedione, acenaphthenedione, 1,2-acenaphthenequinone, 1,2-acenaphthenedione, acenaphthaquinone, acenaphthylene-1,2-quinone, acenaphthylenequinone, acenaphthodione
6724
CHEBI:15342
acenaphthylene-1,2-dione
C1=CC2=C3C(=C1)C(=O)C(=O)C3=CC=C2
82-86-0
C12H6O2
182.18
MFCD00003805
AFPRJLBZLPBTPZ-UHFFFAOYSA-N
acenaphthenequinone, acenaphthoquinone, 1,2-acenaphthylenedione, acenaphthenedione, 1,2-acenaphthenequinone, 1,2-acenaphthenedione, acenaphthaquinone, acenaphthylene-1,2-quinone, acenaphthylenequinone, acenaphthodione
6724
CHEBI:15342
acenaphthylene-1,2-dione
C1=CC2=C3C(=C1)C(=O)C(=O)C3=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 257.0°C to 263.0°C
Color Ochre
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 07, 744
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,benzene,soluble in ligroin
Formula Weight 182.18
Percent Purity 95%
Physical Form Powder or Granules
Chemical Name or Material Acenaphthenequinone
Show More Show Less
compliance-icons
Product Identifier
  • Acenaphthenequinone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'