missing translation for 'onlineSavingsMsg'
Learn More

ACEA, Tocris Bioscience™

Potent and highly selective CB1 agonist

Brand:  Tocris (Bio-Techne) 13195

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 5mg

 View more versions of this product

Product Code. 13449133

  • £83.79 / 5mg

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and highly selective CB1 receptor agonist (Ki = 1.4 nM). Displays >1400-fold selectivity over CB2 receptors. Active in vivo.
Specifications

Chemical Identifiers

220556-69-4
365.986
arachidonyl-2 inverted exclamation marka-chloroethylamide hydrate, 5e,8e,11e,14e-n-2-chloroethyl icosa-5,8,11,14-tetraenamide
(5E,8E,11E,14E)-N-(2-chloroethyl)icosa-5,8,11,14-tetraenamide
C22H36ClNO
SCJNCDSAIRBRIA-CGRWFSSPSA-N
5353377
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCCl

Specifications

5mg
220556-69-4
C22H36ClNO
SCJNCDSAIRBRIA-CGRWFSSPSA-N
(5E,8E,11E,14E)-N-(2-chloroethyl)icosa-5,8,11,14-tetraenamide
5353377
ACEA
>98%
arachidonyl-2 inverted exclamation marka-chloroethylamide hydrate, 5e,8e,11e,14e-n-2-chloroethyl icosa-5,8,11,14-tetraenamide
CCCCCC=CCC=CCC=CCC=CCCCC(=O)NCCCl
365.986
365.99
Videos
Documents