missing translation for 'onlineSavingsMsg'
Learn More

Ansell AccuTech™ 91-325 Series Natural Rubber Latex Splash Gloves

Product Code. 11745773 Shop All Ansell Products
missing translation for 'orderingAttributeHoverText'
Size:
6
6.5
7
7.5
8
8.5
9
missing translation for 'unitSize'
200 pairs
This item is not returnable. View return policy

Product Code. 11745773

missing translation for 'mfr': Ansell 91325065

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provides optimal solutions for controlled and sterile environments, respectively. Ansell Edmont™ AccuTech™ 91-325 Series Natural Rubber Latex Splash Gloves are made with a specific natural rubber formulation that offers a high level of comfortable, sensitive, precise handling.

  • With textured fingertips
  • Excellent technical properties
  • Low allergenic content minimizes the risk of latex-associated allergies for the wearer
  • Ideal for biotechnologies, medical device manufacturing, laboratory analysis, pharmaceuticals and the flat panel industry
TRUSTED_SUSTAINABILITY

Specifications

Cleanroom Class Class 100 (ISO 5)
Color Natural
Latex Free No
Material Latex
Product Type Cleanroom Gloves
Size 6.5
Thickness (Metric) 9.8 mm
For Use With (Application) Biotechnologies, analytics testing, blistering operations, pharmaceuticals
Glove Cuff Color Natural
Glove Cuff Style Straight
Quantity 200 Pairs
Series 91-325
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.