Learn More
Fisherbrand™ accuSolo Clinical Pack
Uses ion-exchange technology to remove ionic contaminants below trace level. Fisher Scientific™ AccuSolo Clinical Pack provides the water quality required for clinical applications.
Brand:  Fisherbrand™
Additional Details : Weight : 1.22000kg
Product Code. 15439719
  • £389.00 / Each
Estimated Shipment: 12-09-2024
to see stock.
Add to basket
Description
Description
  • Removes ionic contaminants
  • Matches water quality required for laboratory applications
Specifications
Specifications
AFS-3/3D
Filter, Ion-exchange