missing translation for 'onlineSavingsMsg'
Learn More

National Diagnostics™ AccuGel™ 29:1 (40%)

Brand:  National Diagnostics™ EC-852-450

Code : LF

Additional Details : CAS Number : 79-06-1 Weight : 0.45000kg Transport : UN number : 3426 Chem class : 6.1 Pack group : III

Quantity 450mL

Product Code. 12331469

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

AccuGel™ 19:1 contains 19g of acrylamide per g of methylene bisacrylamide (MBA) cross-linker. AccuGel™ 29:1 contains 29g of acrylamide per g of MBA. These solutions are ideal for the separation of double stranded DNA fragments. AccuGel™ 19:1 is best suited for the range of molecular sizes less than 100bp, while AccuGel™ 29:1 is best suited for fragments between 100bp and 1,000bp. With the addition of urea, AccuGel™ solutions will also formulate superior sequencing gels.
  • Pre-mixed acrylamide: bis-acrylamide solutions
  • Stabilised for a two year shelf life
  • Consistently crystal clear gels AccuGel™ 19:1 and AccuGel™ 29:1 are ready-to-use monomer solutions for the preparation of electrophoresis gels for DNA or RNA.
Specifications

Specifications

Acrylamide, bisacrylamide
Free
450mL
Free
Free
Videos
Documentation

Technical Documents