missing translation for 'onlineSavingsMsg'
Learn More

National Diagnostics™ AccuGel™ 19:1 (40%)

Brand:  National Diagnostics™ EC-850-1

Code : LF

Additional Details : Weight : 1.00000kg Transport : UN number : 3426 Chem class : 6.1 Pack group : III


Product Code. 12301469

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

AccuGel™ 19:1 contains 19g of acrylamide per g of methylene bisacrylamide (MBA) cross-linker. AccuGel™ 29:1 contains 29g of acrylamide per g of MBA. These solutions are ideal for the separation of double stranded DNA fragments. AccuGel™ 19:1 is best suited for the range of molecular sizes less than 100bp, while AccuGel™ 29:1 is best suited for fragments between 100bp and 1,000bp. With the addition of urea, AccuGel™ solutions will also formulate superior sequencing gels.
  • Pre-mixed acrylamide: bis-acrylamide solutions
  • Stabilised for a two year shelf life
  • Consistently crystal clear gels AccuGel™ 19:1 and AccuGel™ 29:1 are ready-to-use monomer solutions for the preparation of electrophoresis gels for DNA or RNA.
Specifications

Specifications

Acrylamide, bisacrylamide
Free
1L
Free
Black
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • AccuGel 19:1 - 40%

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1

Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H361-Suspected of damaging fertility or the unborn child.
  • H372-Causes damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P306+P360-IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains¬†: Acrylamide

Documentation

Technical Documents