missing translation for 'onlineSavingsMsg'
Learn More

Fisher Scientific™ Accu500 Water System

Exceeds ASTM, CLSI, CAP, and ISO Type I water standards. Fisher Scientific™ Accu500 Water System is a versatile purification system that can be used in a number of ways.

Brand:  Fisher Scientific™ RD0PP30UEK

UNSPSC : 41104200

Additional Details : Weight : 48.10000kg

 View more versions of this product

Product Code. 15439709

  • £6760.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Description

  • The fully automated control system contains pre-rinse, a ‘flush’ feature preventing RO membrane fouling, RO membrane and cartridge life monitoring as well as many other programs for convenient operation
  • The pre-rinsing program starts automatically once a new RO membrane is installed so there is no need for user to set additional cleaning procedures
  • Two sets of dual-column purification cartridges work sequentially to ensure thorough removal of ions and organics in water
  • Automatic hourly system recirculation prevents bacterial growth
  • Resistivity is measured by a highly sensitive resistivity sensor
  • Electrical and water compartments are completely segregated, thus safer to operate
  • User can set water dispensing time for easy water collection
  • Both RO and ultrapure water can be dispensed from the water system
  • The system can be fitted with a 30L or 60L PE tank with a liquid level sensor and a tank vent filter with CO² scavenger to protect purified water from atmospheric degradation
  • A 0.2μm filter or ultrafiltration filter (optional) removes final contaminants
  • Independent ultrapure water dispenser
  • The remote dispenser has a built-in high precision resistivity sensor with cell constant of 0.01cm-¹ to measure the quality of ultrapure water just prior to the dispensing point
  • Volumetric dispensing and calibration
  • The handle is 360 degree-rotatable and detachable from its support for effortless and instant dispensing with maximum flexibility
Applications:
Ultrapure and RO water can be used in many areas. Some typical applications include:

Ultrapure water
  • HPLC mobile phase preparation
  • Preparation of reagent blank solutions
  • As sample diluent for GC, HPLC, AA, ICP-MS and other analytical techniques
  • Preparation of buffers and culture media for mammalian cell culture
  • Preparation of molecular biology reagents, etc.
RO water
  • Glassware cleaning
  • Washing machine for glassware
  • Waterbath water
  • Autoclaving
Description & Specifications

Specifications

Municipal tap water, TDS <1000 ppm (2000 μS/cm)
<20μS/cm
<1/mL with 0.2μm final filter or terminal ultrafiltration cartridge
48cm
5 to 45°C
Accu500
30L/hr
18.2 MΩ·cm
<0.001Eu/mL with a terminal ultrafiltration cartridge
1 to 6 bar
30cm
51cm
1.5L/min
Accu500 UV Ultrapure water system with dispenser
Documentation Special Offers

Special Offers