Learn More
Thermo Scientific™ Acclaim™ SEC-1000 Size Exclusion Chromatography HPLC Columns
Separate water-soluble polymers and oligomers with the Acclaim family of resin-based, high-performance, size-exclusion chromatography columns.
Brand: Thermo Scientific™ 079724
Code : 15
Additional Details : Weight : 0.10000kg
Description
Separate water-soluble polymers and oligomers with Thermo Scientific™ Acclaim™ SEC-1000 Size Exclusion HPLC Columns. These resin-based, high-performance, size-exclusion chromatography columns cover molecular weight range: 7μm, 1000Å. The combination of a proprietary mono-dispersed multi-pore hydrophilic resin and small particle size provides high-resolution and good linearity of the calibration curve with no inflection points for accurate, reliable analysis of a variety of water-soluble polymers.
The Acclaim SEC-1000 LC column is calibrated from 1,000 to 1,000,000 Daltons and can characterize high molecular weight polymers.High-Performance Analysis of Water-Soluble Polymers
- High-resolution analysis at reduced solvent consumption in available 4.6 x 300mm dimension
- Stable surface bonding leads to low column bleed and enhances compatibility with UV, RI, MS, ELSD and Thermo Scientific Dionex™ Corona™ Charged Aerosol Detectors
Specifications
Analytical Column | |
0.35 mL/min | |
600 psi (40 bar) | |
7 μm | |
1000 Å | |
Proprietary | |
Spherical, Fully Porous Polymer | |
Yes | |
Analytical Column |
4.6 mm | |
300 mm | |
Spherical | |
2 to 12 | |
60°C | |
Acclaim | |
Size Exclusion | |
Size Exclusion | |
Bio-LC |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.