missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acclaim™ SEC-300 LC Columns

Separate water-soluble polymers and oligomers with the Acclaim family of resin-based, high-performance, size-exclusion chromatography columns.

Brand:  Thermo Scientific™ 079726

Code : 15

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15128754

  • £1290.00 / Each


Estimated Shipment
19-06-2018
Add to basket

Description

Description

The Acclaim SEC-300 column is calibrated from 100 to 50,000 Daltons and offers good resolution for lower molecular weight polymers or oligomers.

High-Performance Analysis of Water-Soluble Polymers
  • High-resolution analysis at reduced solvent consumption in available 4.6 x 300mm dimension
  • Stable surface bonding leads to low column bleed and enhances compatibility with UV, RI, MS, ELSD and Thermo Scientific Dionex™ Corona™ Charged Aerosol Detectors
Specifications

Specifications

Analytical Column
7.8mm
1.00mL/min
700 psi
Spherical
2 to 12
60°C
Proprietary
Size Exclusion
Yes
150mm
Polymer, Spherical Fully Porous
5μm
300Å
N/A
Videos

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.