Learn More
Thermo Scientific™ PepMap™ 100 C8 HPLC Columns
Use this nanocolumn for high-resolution analyses of tryptic, natural, and synthetic peptides.
£318.24 - £547.74
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
11382153
|
Thermo Scientific™
161185 |
3 μm | 0.075 mm | 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11372163
|
Thermo Scientific™
161194 |
5 μm | 0.3 mm | 5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Choose Thermo Scientific™ Acclaim™ PepMap™ 100 C8 LC Columns when separating very hydrophobic peptides (nontryptic peptides) as an alternative to the PepMap™ C18 column.
This column provides high-resolution, high-sensitivity protein identification for proteomics and biomarker discovery and verification, and is ideal for:
High loading capcity
Acclaim PepMap columns are exceptionally suitable for the analysis of low abundant peptides often found in complex proteomics samples.
The PepMap 100 C8 LC Columns is available in 150mm length with a choice of 75, 300, 1000, and classic 75μm I.D. to allow tuning for throughput, sensitivity and amount of sample available.
Superior protein and peptide separations
Trifluoroacetic acid (TFA)-free LC/MS minimizes suppression effects for enhanced MS sensitivity
Outstanding separation efficiency
Superior resolution and recovery
Easy installation with Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fittings
Columns are available with nanoViper Fingertight Fittings
Requires no tools
Provides virtually zero-dead-volume connections
Withstands pressures up to a 1000 bar
About Nano, capillary, and micro Acclaim PepMap columns
Nano (75μm), capillary (300μm) and micro (1000μm) columns are ideal for protein and peptide analyses. Sample amount is often limited and smaller I.D. columns can increase in sensitivity in sample-limited situations. While nano LC columns are often used in proteomics, the concentrating effects of capillary and micro columns are beneficial for metabolomics or for facilitating LC-MS interfacing. Thermo Scientific Acclaim PepMap columns are offered in nano, capillary and micro column in a variety of chemistries for peptides and protein analysis.
Specifications
100 Å | |
Reversed Phase | |
Stainless Steel |