missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Titanium-Dioxide Nano-Trap Columns
Titanium-Dioxide Nano-Trap Columns support the enrichment of phosphopeptides.
£202.98 - £258.06
Specifications
Column Format | Trap Column |
---|---|
Length (Metric) | 10 mm |
Max. Pressure | 7250 psi (500 bar) |
Particle Size | 5 μm |
Stationary Phase | Titanium Dioxide |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
11352253
|
Thermo Scientific™
164215 |
5 μm | 0.2 mm | 10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11302253
|
Thermo Scientific™
164205 |
5 μm | 0.1 mm | 10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Support the enrichment of phosphopeptides with Thermo Scientific™ Titanium-Dioxide Nano-Trap Columns.
Specifications
Trap Column | |
7250 psi (500 bar) | |
Titanium Dioxide | |
Titanium Dioxide | |
Reversed Phase | |
Trap Column |
10 mm | |
5 μm | |
nanoLC | |
Reversed Phase | |
Fused Silica |