missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Titanium-Dioxide Nano-Trap Columns
Titanium-Dioxide Nano-Trap Columns support the enrichment of phosphopeptides.
Brand: Thermo Scientific™ 164205
Code : 16
Additional Details : Weight : 0.10000kg
Description
Support the enrichment of phosphopeptides with Thermo Scientific™ Titanium-Dioxide Nano-Trap Columns.
Specifications
Trap Column | |
10 mm | |
5 μm | |
nanoLC | |
Reversed Phase | |
Fused Silica |
0.1 mm | |
7250 psi (500 bar) | |
Titanium Dioxide | |
Titanium Dioxide | |
Reversed Phase | |
Trap Column |