Learn More
Thermo Scientific™ Acclaim™ 5μm PepMap™ 100 Fusica™ Microbore Columns
Feature high loading capacity for exceptional suitability in the analysis of low abundant peptides in complex proteomics samples.
Brand: Thermo Scientific™ 160287
Code : 16
Additional Details : Weight : 0.10000kg
Description
Feature high loading capacity for exceptional suitability in the analysis of low abundant peptides in complex proteomics samples with Thermo Scientific™ Acclaim 5μm PepMap 100 Fusica™ Microbore Columns.
- High resolution in protein identification, biomarker discovery, and systems biology
- Particular loadability allows sensitivity in LC/MS
- Designed for TFA-free LC-MS, minimizing ion-suppression effects
- Optimized for coupling to ESI/MS and MALDI-MS
- Highest column-to-column reproducibility
- Easy-to-use, cutting-edge miniaturized HPLC
- Inside diameter 1.0mm
Specifications
Capillary Column | |
40 µL/min | |
250 mm | |
2 to 8 | |
450 m²/g | |
C18 | |
15% | |
Reversed Phase |
1 mm | |
LC/MS, ESI-MS, MALDI-MS | |
5 μm | |
100 Å | |
L1 | |
Silica | |
Peptide Analysis | |
Stainless Steel |