missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Accessories for XX6702500 FilterJet Solvent Dispenser
Accessories for filter holders for sample preparation.
Brand: Merck XX4002500
Code : 72
Additional Details : Weight : 0.18048kg
Description
Accessories for filter holders for sample preparation.
Specifications
Filterjet Solvent Dispenser | |
Filter Holder | |
2.5 cm |
Stainless Steel Filter Holder | |
0.98 in. |