Learn More
Thermo Scientific™ Accessories for Thermo Scientific™ Refrigerated and Heated Bath Circulators
Enhance the operation of Refrigerated and Heated Bath Circulators with accessories that include bridges, platforms, cooling coils and connectivity solutions.
Brand: Thermo Scientific™ 1600018
Code : 50
Additional Details : Weight : 0.10000kg
Description
Enhance the operation of Refrigerated and Heated Bath Circulators with Thermo Scientific™ Accessories which include bridges, platforms, cooling coils and connectivity solutions.
- Warranty varies by country, please contact us for details
Specifications
Adjustable bridge in widths between 300 and 800 mm, for SC, AC and PC immersion circulators | |
Thermo Scientific™ Refrigerated and Heated Bath Circulators | |
Bridge |