Learn More
Thermo Scientific™ Accessories for Thermo Scientific™ Refrigerated and Heated Bath Circulators
Enhance the operation of Refrigerated and Heated Bath Circulators with accessories that include bridges, platforms, cooling coils and connectivity solutions.
Brand: Thermo Scientific™ 1600078
Code : A0
Additional Details : Weight : 0.80000kg
Description
Enhance the operation of Refrigerated and Heated Bath Circulators with Thermo Scientific™ Accessories which include bridges, platforms, cooling coils and connectivity solutions.
- Warranty varies by country, please contact us for details
Specifications
Stainless-steel bridge to hold SC controller on the W13, W15, W26, W45, W46 baths | |
Thermo Scientific™ Refrigerated and Heated Bath Circulators | |
Bridge |