Learn More
Thermo Scientific™ Accessories and Replacement Parts for MaxiMix™ III Vortex Mixer
Optimize flexibility of your mixer with this selection of accessories and replacement parts.
Brand: Thermo Scientific™ PT500X9A
Code : NEW
Additional Details : Weight : 1.20000kg
Description
Utilize these Thermo Scientific™ Accessories and Replacement Parts for MaxiMix™ III Vortex Mixer to optimize flexibility of the machine. Options include a utility tray, flask holder and universal holder for a variety of flasks, beakers, bottles, test tub racks
Specifications
Universal Holder | |
Universal holder for a variety of flasks, beakers, bottles, test tub racks. Two soft rubber bars hold vessels in place. | |
MaxiMix™ III Vortex Mixer |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.