Learn More
Thermo Scientific™ Electrode Storage Bottles for 12mm pH electrode, 3/pk
Store and protect your pH electrodes with these accessories, including storage bottles and electrode bulb guards.
Brand:  Thermo Scientific™ 910003
Code : 04
Additional Details : Weight : 0.10000kg
Product Code. 10614131
  • £56.71 / Pack of 3
In Stock
to see stock.
Add to basket
Description
Description
Store and protect your pH electrodes with the Thermo Scientific™ Accessories for Orion™ pH Electrodes. Storage bottles protect and condition electrodes when not in use. Bulb guard protects the pH-sensing bulb of glass pH electrodes.
Electrode Storage Bottles
  • Can be used with any 6, 8 or 12mm pH electrodes
Electrode Bulb Guards
  • Designed for glass body pH electrodes
  • Use with standard or Thermo Scientific™ Orion™ Sure-Flow™ pH electrodes — one end of the bulb guard fits over standard electrodes while the other end fits over Sure-Flow electrodes without interfering with the Sure-Flow flushing action