Learn More
Cytiva Whatman™ Accessories for Mini-UniPrep™ Syringeless Filters
Used with Mini-UniPrep syringeless plunger filter units for fast and easy HPLC/UHPLC sample preparation. Whatman™ Compressors for Mini-UniPrep™ Syringeless Filters process one or multiple filter vials simultaneously for
£16.15 - £236.00
Specifications
For Use With (Application) | Sample filtration/preparation for HPLC/UHPLC |
---|---|
Housing Material | Polypropylene |
Quantity | 1 Each |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15244548
|
Cytiva
MUPG2MCWT8 |
Vial tray for the G2 Multi-compressor (mfr. no. MUPG2MCPWC8) |
£16.30
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
15289623
|
Cytiva
MUPG2MCPWC8 |
Multi-Sample Compressor Unit; Eight positions |
£236.00
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
15530143
|
Cytiva
MUPG2HCPWC1 |
Hand compressor, Single position (Promo) |
£16.15
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
11318724
|
Cytiva
CR0000006 |
Multi-sample Hand Compressor; Six positions |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Whatman Mini-UniPrep Syringeless Filters integrate an autosampler vial, filtration membrane, plunger, and cap/septa into one consumable product. Compressors activate the plunger to filter the sample through the membrane without hand stress.
- Compatible with Mini-UniPrep and/or Mini-UniPrep G2 syringeless filter vials
- Multi-chamber compressors process multiple vials simultaneously for stream-lined filtration
- Mini-UniPrep filter vials prepare samples 3x faster than standard syringe filters, and can be loaded directly into an autosampler
- Options include: 1-, 6- or 8-position hand or handle compressors (additional 8-position trays also available)
Specifications
Sample filtration/preparation for HPLC/UHPLC | |
Polypropylene | |
1 Each |