missing translation for 'onlineSavingsMsg'
Learn More

Acacia, Total ash <4%, Thermo Scientific Chemicals

Catalog Number p-7050324
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
9000-01-5
MFCD00081264
Gum Arabic
This item is not returnable. View return policy
9000-01-5
MFCD00081264
Gum Arabic

Mixture of polysaccharides and glycoproteins used in the food industry as a stabilizer

It is used in textiles, ceramics, lithography, cosmetics and pharmaceuticals. In the food industry, acacia is mainly used as confectionery, bakery, dairy, drink, and as a microencapsulating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Density 1.4 g/mL
Mesh Size −100 Mesh
Odor Uncharacteristic
Quantity 500 g
Merck Index 14,14
Solubility Information Water soluble. Aqueous solution is acidic to litmus.
Physical Form Powder
Chemical Name or Material Acacia, Total ash <4%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'