missing translation for 'onlineSavingsMsg'
Learn More

AC 261066, Tocris Bioscience™

RARβ2 agonist

Brand:  Tocris (Bio-Techne) 404650

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13444034

  • £586.53 / 50mg

Estimated Shipment date
from Supplier 20-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent RARβ2 agonist (pEC50 = 8.1). Selective over RARβ1, RARα and RARγ (pEC50 values are 6.4, 6.2 and 6.3 respectively). More potent and orally available than AC 55649 (Cat. No. 2436).
Specifications

Chemical Identifiers

870773-76-5
353.408
unii-lgf3f89bzp, lgf3f89bzp, 4-4-2-butoxyethoxy--5-methyl-2-thiazolyl-2-fluorobenzoic acid, benzoic acid, 4-4-2-butoxyethoxy-5-methyl-2-thiazolyl-2-fluoro, 4-4-2-butoxyethoxy-5-methyl-1,3-thiazol-2-yl-2-fluorobenzoic acid, d09rrf, 4-4-2-butoxyethoxy-5-methylthiazol-2-yl-2-fluorobenzoic acid
4-[4-(2-butoxyethoxy)-5-methyl-1,3-thiazol-2-yl]-2-fluorobenzoic acid
C17H20FNO4S
HSAOETBFVAWNRP-UHFFFAOYSA-N
11530459
CCCCOCCOC1=C(SC(=N1)C2=CC(=C(C=C2)C(=O)O)F)C

Specifications

50mg
870773-76-5
unii-lgf3f89bzp, lgf3f89bzp, 4-4-2-butoxyethoxy--5-methyl-2-thiazolyl-2-fluorobenzoic acid, benzoic acid, 4-4-2-butoxyethoxy-5-methyl-2-thiazolyl-2-fluoro, 4-4-2-butoxyethoxy-5-methyl-1,3-thiazol-2-yl-2-fluorobenzoic acid, d09rrf, 4-4-2-butoxyethoxy-5-methylthiazol-2-yl-2-fluorobenzoic acid
CCCCOCCOC1=C(SC(=N1)C2=CC(=C(C=C2)C(=O)O)F)C
353.408
353.41
AC 261066
C17H20FNO4S
HSAOETBFVAWNRP-UHFFFAOYSA-N
4-[4-(2-butoxyethoxy)-5-methyl-1,3-thiazol-2-yl]-2-fluorobenzoic acid
11530459
>98%
Videos
Documents