Learn More
Tocris Bioscience™ AC 186
Potent and selective ERβ agonist; neuroprotective
POA
Specifications
Product Type | AC 186 |
---|---|
Description | AC 186 |
Molecular Weight (g/mol) | 306.32 |
Description
Potent and selective ERβ agonist (EC50 values are 6 and 5000 nM for ERβ and ERα respectively). Exhibits gender selective neuroprotective effects in a male rat model of Parkinson's disease. Decreases Aβ levels in combination with ACP-105 in a rat model of Alzheimer's disease.Specifications
AC 186 | |
AC 186 | |
306.32 |
For Research Use Only