Learn More
Tocris Bioscience™ ABT 724 trihydrochloride
Potent and selective D4 partial agonist; proerectile
Brand:  Tocris Bioscience™ 2214/10
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13450814
  • £127.89 / 10mg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
Potent dopamine D4 receptor partial agonist (EC50 = 12.4 nM; 61% efficacy vs. dopamine). Has no agonist activity at D2 receptors (EC50 > 10 μM). Selective in rats in vivo; produces penile erection following i.c.v. administration; increases intracavernosal pressure and potentiates the proerectile effects of sildenafil following s.c. administration. Displays minimal side effects.
Specifications
Specifications
ABT 724 trihydrochloride
>99
10mg
ABT 724 trihydrochloride
402.75