missing translation for 'onlineSavingsMsg'
Learn More

ABsolute™ qPCR Master Mixes, SYBR Green-Based Detection

Thermo Scientific™ ABsolute qPCR Master Mixes offer sensitivity and reproducibility for real-time PCR applications.

Brand:  Thermo Scientific ABgene AB1219B

Code : LE

Additional Details : Weight : 0.09000kg

 View more versions of this product

Product Code. 11832063

  • £1348.05 / 1600 reactions


EU Stock 5
Add to basket

Description

Description

Highlights

  • High Sensitivity & Specificity -The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
  • Optimised buffer containing SYBR Green I
  • Reproducibility -All master mixes undergo extensive QC to ensure batch to batch consistency
  • Broad dynamic range
Specifications

Specifications

heated to 95°C for 15 minutes
ABsolute qPCR Buffer (Thermo-Start DNA polymerase, dNTPs, MgCl2, SYBR Green I, ROX™ passive reference dye (only in ROX mixes), Fluorescein (only in Fluorescein mix)); Separate MgCl2; Separate ROX vial (only in kits indicated)
-20°C Avoid freeze/thaw cycles
ABsolute QPCR SYBR Green Fluorescein Mix
800 x 50µL rxns
Videos

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.