Learn More
Thermo Scientific™ ABsolute Blue QPCR Mix, SYBR Green, low ROX
Incorporates an inert blue dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof.
Brand: Thermo Scientific™ AB4323A
547.20 GBP valid until 2024-12-31
Use promo code "21881" to get your promotional price.
Code : A0
Additional Details : CAS Number : 7732-18-5 Weight : 0.25000kg
Description
Thermo Scientific ABsolute Blue qPCR SYBR Green Mixes are optimized 2X master mixes for SYBR Green chemistry. The master mixes incorporate an inert blue dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof. Absolute Blue qPCR Mixes include Thermo-Start Taq hot start enzyme for increased sensitivity.
- High performance - ABsolute sensitivity, consistency & convenience
- Enhanced contrast and reduced errors - easily identify correctly aliquotted reaction mix in all plates and tubes
- Universal - compatible with all major qPCR platforms
- Adaptable - available with ROX or Fluorescein passive reference dyes
Specifications
Contains: • 1 x 5 mL ABsolute Blue QPCR SYBR Green Low ROX Mix (2X) • 100 μL MgCl2 (1M) Sufficient for 400 x 25 μL reactions. Store at -20°C |
|
Tube | |
qPCR | |
Standard | |
ABsolute Blue | |
400 Reactions of 25 μL Each | |
qPCR Cyclers | |
400 Reactions | |
5 mL |
SYBR | |
High | |
Taq DNA Polymerase | |
7500 System, QuantStudio™, ViiA™ 7 System, Stratagene Mx4000, Stratagene Mx3000P, Stratagene Mx3005P | |
Real Time PCR SYBR Master Mix | |
Gene Expression, Genotyping, siRNA Validation | |
2X | |
DNA, dsDNA |
For Research Use Only. Not for use in diagnostic procedures.