(±)-Abscisic Acid 98%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 133485000

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 14375-45-2 Weight: 0.05050kg


Packaging Glass bottle
Quantity 500mg

Product Code. 10357751

Quantity Price
1 £ 101.6 / 500mg
In Stock 6
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material (±)-Abscisic acid
Name Note 98%
CAS 14375-45-2
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C15H20O4
Formula Weight 264.32
MDL Number MFCD00075619
Synonym (2-cis, 4-trans)-5-(1-Hydroxy-2, 6

Specification

Formula Weight 264.32
Quantity 500mg
Merck Index 15, 12
CAS Min % 97.5
CAS 14375-45-2
Assay Percent Range 98%
MDL Number MFCD00075619
Sublimation Point 0120.0°C
Packaging Glass bottle
Molecular Formula C15H20O4
Health Hazard 1 LIGHT SENSITIVE
Name Note 98%
CAS Max % 100.0
Synonym (2-cis, 4-trans)-5-(1-Hydroxy-2, 6
TSCA TSCA
Chemical Name or Material (±)-Abscisic acid
Documentation

Technical documents

Safety and Handling

LIGHT SENSITIVE