Learn More
Abnova™ Tnfrsf11a (Mouse) ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Tnfrsf11a
Brand:  Abnova™ KA2007.1Kit
Additional Details : Weight : 0.02000kg
Product Code. 16036995
  • £527.81 / 1 set
Estimated Shipment
19-06-2020
Add to basket
Specifications
Specifications
Biotin
Tnfrsf11a (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Tnfrsf11a.
Colorimetric
96-well Antibody coated microwell, lyophilized recombinant Mouse Tnfrsf11a standard 10ng x 2 vials, 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Mouse Tnfrsf11a Antibody, 12mL Antibody Diluent Buffer, 130μL Avidi
TNFRSF11A (Mouse) ELISA kit
Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate
100μL
Mouse
<2pg/mL
Quantification
Tnfrsf11a (Mouse) ELISA Kit
1 Kit
Store the kit at-20°C.