Learn More
Abnova™ Insulin (Human) ELISA Kit
For quantitative measurement Insulin in Human Serum or Plasma
Brand:  Abnova™ KA3810.1Kit
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16018245
  • £334.56 / 1 set
Estimated Shipment
22-06-2020
Add to basket
Specifications
Specifications
0 to 140μIU/mL
Insulin (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human insulin.
Quantification
Insulin (Human) ELISA kit
Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysates
Store the kit at 4°C.
Human
Colorimetric
Insulin (Human) ELISA Kit
1 Kit
50μL