missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Insulin Ab ELISA Kit

indirect solid phase ELISA for the quantitative measurement of IgG class autoantibodies against Bovine, Porcine and recombinant Human Insulin in Human Serum or Plasma

Brand:  Abnova™ KA1107.1Kit

Additional Details : Weight : 0.02000kg

Product Code. 16096545

  • £241.05 / 1 set


Estimated Shipment
18-06-2020
Add to basket

Specifications

Specifications

6.3 to 100U/mL
Insulin Ab ELISA Kit is an indirect solid phase ELISA for the quantitative measurement of IgG class autoantibodies against bovine, porcine and recombinant human insulin in human serum or plasma.
Colorimetric
6 Vials, 1.5mL calibrator with IgG class Anti-Insulin Antibodies, 2 vials x 1.5mL Anti-Insulin controls in a Serum/Buffer Matrix, 1 vial x 20mL Sample Buffer, 1 vial x 15mL Enzyme Conjugate solution, 1 vial x 15mL TMB Substrate solution,, 1 vial x 15mL stop solution, 1 96-well plate, 1 vial x 20mL wash solution
96 Assays
Store the kit at 4°C.
10μL
HRP
0.5U/mL
Quantification
Insulin Ab ELISA Kit
Plasma, Serum
96 Assays
Videos
Documents