missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Fluorescent Dye AMCA Phalloidin

Product Code. 16149370
Click to view available options
Quantity:
300 Reactions
Unit Size:
300 reactions
This item is not returnable. View return policy

Product Code. 16149370

Brand: Abnova™ U0308.300Reactions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorescence Dye AMCA with Phalloidin conjugate

Ideal for labeling via Phalloidin for fluorescence imaging and other fluorescence-based biochemical analysis.

  • Excitation (Max): 353nm
  • Emission (Max): 442nm
  • Solubility: DMSO
  • Storage Buffer: DMSO

Application: Conjugation

Recommended Storage: Store in desiccated environment at -20°C and avoid from light.

Specifications

Description Fluorescence Dye AMCA with Phalloidin conjugate.
For Use With (Application) Conjugation
Quantity 300 Reactions
Product Type Fluorescent Dye AMCA Phalloidin
Sufficient For 300 Reactions
Storage Buffer DMSO
Content And Storage Store the product in desiccated environment at -20°C and avoid from light.
Label or Dye Fluorescent Dye AMCA Phalloidin
Emission 442
Excitation 353

For research use only (RUO).

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.