Learn More
Abnova™ Fluorescent Dye 790-I Phalloidin
Description
The fluorescence emission maximum of Fluorescent Dye 790-I around 810nm is well separated from commonly used far-red fluorophores, including Fluorescent Dye 647-I, Fluorescent Dye 680-I or allophycocyanin (APC), facilitating the rapid growing NIR biological analysis. Fluorescent Dye 790-I is one of the brightest NIR fluorescent labeling dyes with fluorescence quantum yield close to 0.1. Its conjugates have been successfully used for NIR fluorescent probe-based in vivo imaging analysis. Ideal for labeling via Phalloidin for fluorescence imaging and other fluorescence-based biochemical analysis.
- Spectral Equivalent: Alexa Fluor 790, IRDye 800
- Molar Extinction Co-efficient: ≥2,50,000 M-1cm-1
- Excitation (Max): 783nm
- Emission (Max): 814nm
- Solubility: DMSO
- Storage Buffer: DMSO
Application: Conjugation, invivo imaging
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescence Dye 790-I with Phalloidin conjugate. The fluorescence emission maximum of Fluorescent Dye 790-I around 810nm is well separated from commonly used far-red fluorophores, including Fluorescent Dye 647-I, Fluorescent Dye 680-I or allophycocyanin (APC), facilitating the rapid growing NIR biological analysis. Fluorescent Dye 790-I is one of the bightest NIR fluorescent labeling dyes with fluorescence quantum yield close to 0.1. Its conjugates have been successfully used for NIR fluorescent probe-based in vivo imaging analysis. |
| For Use With (Application) | Conjugation; in vivo Imaging |
| Quantity | 300 Reactions |
| Product Type | Fluorescent Dye 790-I Phalloidin |
| Sufficient For | 300 Reactions |
| Equivalence | Alexa Fluor 790, IRDye 800 |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 790-I Phalloidin |
| Emission | 814 |
| Show More |
For research use only (RUO).
Your input is important to us. Please complete this form to provide feedback related to the content on this product.