missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Fluorescent Dye 647-I Azide
Click to view available options
:
Quantity: 1mg
Description
The slight red shift in absorption spectrum makes Fluorescent Dye 647-I Azide an optimal match to filters designed Cy5 dyes. Besides, their fluorescence is pH-independent from pH 3 to 11. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis
- Spectral Equivalent: Alexa Fluor 647, Cy5, DyLight 650
- Purity: >90%
- Molar Extinction Co-efficient: >2,40,000 M-1cm-1
- Excitation (Max): 649nm
- Emission (Max): 664nm
- Solubility: DMSO, water
- Storage Buffer: DMSO
Application: Conjugation, Flow Cytometry
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Water-soluble fluorescent dye reacting with alkyne groups. The slight red shift in absorption spectrum makes Fluorescent Dye 647-I Azide an optimal match to filters designed Cy5 dyes.Besides, their fluorescence is pH-independent from pH 3 to 11. |
| For Use With (Application) | Conjugation; Flow Cytometry |
| Quantity | 1 mg |
| Product Type | Fluorescent Dye 647-I Azide |
| Purity | >90% |
| Equivalence | Alexa Fluor 647, Cy5, DyLight 650 |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 647-I Azide |
| Emission | 664 |
| Show More |
For research use only (RUO).
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction