missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Fluorescent Dye 645-A Amine
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
- Chemical Name: 2-((1E,3E,5E)-5-(1-(3-ammoniopropyl)-3,3-dimethyl-5-sulfonatoindolin-2-ylidene)penta-1,3-dienyl)-1-ethyl-3,3-dimethyl-3H-indolium-5-sulfonate
- Chemical Formula: C30H37N3O6S2
- Mol. Weight: 599.76g/mol
- Purity: ≥95%
- Molar Extinction Co-efficient: 2,50,000 M-1cm-1 (CH3COH)
- Excitation (Max): 643nm (CH3OH)
- Emission (Max): 662nm (CH3OH)
- Recommended Excitation: Argon-Krypton ion laser 647nm, Red diode laser 635nm
- Recommended Filter Sets: Cy5, APC
- Solubility: Methanol, DMSO, DMF, water, acetonitrile
Application: Conjugation
Recommended Storage: Store in the dark at -20°C.
Specifications
Specifications
| Description | Fluorescent dye derived from Cyanine. Water soluble and reacts toward activated carboxylic acid at pH 7-9 inserting a negative charge in the labeled molecule. |
| Formulation | Derived from Cyanine |
| For Use With (Application) | Conjugation |
| Quantity | 5 mg |
| Product Type | Fluorescent Dye 645-A Amine |
| pH Range | 7 to 9 |
| Molecular Weight (g/mol) | 599.76g/mol |
| Light Source | Argon-Krypton ion laser 647nm, Red diode laser 635nm |
| Filter Type | APC, Cy5 |
| Purity | ≥95% |
| Show More |
For research use only (RUO).
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction