Learn More
Abnova™ 5-Color FISH Probe (chromosome 13, 18, 21, X and Y)
Labeled FISH probes for identification of prenatal chromosomal abberations using Fluorescent In Situ Hybridization Technique.
Brand: Abnova™ FM0003.100uL
This item is not returnable.
View return policy
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Specifications
Store at 4°C in the dark. | |
Fluorescent In Situ Hybridization (Cell) | |
CEN13q, CEN18q, CEN21q, CENXp, CENYq | |
100 μL |
Cy5, DEAC, FITC, R6G, Texas Red | |
13q12.11, 18q11.2, 21q22.13, Xp11.23, Yq11.221 | |
5-Color FISH Probe (chromosome 13, 18, 21, X & Y) |