missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ AARS Recombinant Mouse Protein, His tag

Expressed from DNA sequence encoding mouse AARS (Met 1-Asn 968) fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 50384M08B5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15323176

  • £83.81 / 5µg


Estimated Shipment
05-07-2018
Add to basket

Description

Description

  • Formulation: 20mM Tris, 500mM NaCl, 10% Glycerol, pH 7.4
Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
234734
≥85% by SDS-PAGE
Mouse
<1 EU/μg
AARS
Other Proteins
AARS Recombinant Mouse Protein
Lyophilized
108.3kd
5μg
Full Length, Recombinant
Alanyl-Trna Synthetasep
Assay Standard
Insect
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.