missing translation for 'onlineSavingsMsg'
Learn More

AACOCF3, Tocris Bioscience™

Phospholipase A2 inhibitor

Brand:  Tocris (Bio-Techne) 146225

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 25mg

 View more versions of this product

Product Code. 13459543

  • £360.37 / 25mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Inhibitor of cytosolic (85 kDa) phospholipase A2. Also inhibits fatty acid amide hydrolase (FAAH, anandamide amidase) in vitro.
Specifications

Chemical Identifiers

149301-79-1
356.473
aacocf3, arachidonyl trifluoromethyl ketone, arachidonyltrifluoromethane, arachidonic acid trifluoromethyl ketone, arachidonyltrifluoromethyl ketone, atfmk, unii-00xiw1cr0f, 6z,9z,12z,15z-1,1,1-trifluorohenicosa-6,9,12,15-tetraen-2-one, 00xiw1cr0f, 1,1,1-trifluoro-6z,9z,12z,15z-heneicosatetraen-2-one
CHEBI:2341
CCCCCC=CCC=CCC=CCC=CCCCC(=O)C(F)(F)F
C21H31F3O
PLWROONZUDKYKG-DOFZRALJSA-N
5280436
(6Z,9Z,12Z,15Z)-1,1,1-trifluorohenicosa-6,9,12,15-tetraen-2-one

Specifications

AACOCF3
149301-79-1
aacocf3, arachidonyl trifluoromethyl ketone, arachidonyltrifluoromethane, arachidonic acid trifluoromethyl ketone, arachidonyltrifluoromethyl ketone, atfmk, unii-00xiw1cr0f, 6z,9z,12z,15z-1,1,1-trifluorohenicosa-6,9,12,15-tetraen-2-one, 00xiw1cr0f, 1,1,1-trifluoro-6z,9z,12z,15z-heneicosatetraen-2-one
CCCCCC=CCC=CCC=CCC=CCCCC(=O)C(F)(F)F
356.473
CHEBI:2341
>98%
25mg
C21H31F3O
PLWROONZUDKYKG-DOFZRALJSA-N
(6Z,9Z,12Z,15Z)-1,1,1-trifluorohenicosa-6,9,12,15-tetraen-2-one
5280436
356.47
Videos