Learn More
AA 29504, Tocris Bioscience™
Positive allosteric modulator of GABAA receptors
Brand:  Tocris (Bio-Techne) 397210
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13443914
  • £183.75 / 10mg
Estimated Shipment date
from Supplier 25-03-2020
Description
Description
Positive allosteric modulator of GABAA receptors. Modulates both α4β3δ-containing extrasynaptic receptors and α1β3γ2S-containing receptors in Xenopus oocytes, displaying a stronger modulatory effect at the former. Reduces motor coordination and exhibits anxiolytic effects in vivo.
Specifications
Chemical Identifiers
945828-50-2
327.428
aa hplc, 2-amino-4-2,4,6-trimethylbenzylamino-phenyl-carbamic acid ethyl ester, ethyl n-2-amino-4-2,4,6-trimethylphenyl methylamino phenyl carbamate, ethyl n-2-amino-4-2,4,6-trimethylphenyl methyl amino phenyl carbamate, n1-ethoxycarbonyl-n4-2,4,6-trimethylpheny lmethyl-1,2,4-triaminobenzene
ethyl N-[2-amino-4-[(2,4,6-trimethylphenyl)methylamino]phenyl]carbamate
C19H25N3O2
SCOOTUMXCXKEAD-UHFFFAOYSA-N
23188887
CCOC(=O)NC1=C(C=C(C=C1)NCC2=C(C=C(C=C2C)C)C)N
Specifications
10mg
945828-50-2
aa hplc, 2-amino-4-2,4,6-trimethylbenzylamino-phenyl-carbamic acid ethyl ester, ethyl n-2-amino-4-2,4,6-trimethylphenyl methylamino phenyl carbamate, ethyl n-2-amino-4-2,4,6-trimethylphenyl methyl amino phenyl carbamate, n1-ethoxycarbonyl-n4-2,4,6-trimethylpheny lmethyl-1,2,4-triaminobenzene
CCOC(=O)NC1=C(C=C(C=C1)NCC2=C(C=C(C=C2C)C)C)N
327.428
327.42
AA 29504
C19H25N3O2
SCOOTUMXCXKEAD-UHFFFAOYSA-N
ethyl N-[2-amino-4-[(2,4,6-trimethylphenyl)methylamino]phenyl]carbamate
23188887
>98%