missing translation for 'onlineSavingsMsg'
Learn More

AA 2414, Tocris Bioscience™

Thromboxane A2 (TP) receptor antagonist

Brand:  Tocris (Bio-Techne) 302510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13442254

  • £113.40 / 10mg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Thromboxane A2 (TP) receptor antagonist. Antiasthmatic agent; inhibits platelet aggregation and bronchoconstriction induced by a TXA2 mimetic in guinea pigs.
Specifications

Chemical Identifiers

112665-43-7
354.446
seratrodast, bronica, abbott 73001, abbott-73001, seratrodast usan:inn, 7-phenyl-7-2,4,5-trimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl heptanoic acid, bronica tn, ccris 8939, +--7-3,5,6-trimethyl-1,4-benzoquinon-2-yl-7-phenylheptanoic acid, seratrodast jan/usan/inn
7-phenyl-7-(2,4,5-trimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl)heptanoic acid
C22H26O4
ZBVKEHDGYSLCCC-UHFFFAOYSA-N
2449
CC1=C(C(=O)C(=C(C1=O)C)C(CCCCCC(=O)O)C2=CC=CC=C2)C

Specifications

AA 2414
112665-43-7
seratrodast, bronica, abbott 73001, abbott-73001, seratrodast usan:inn, 7-phenyl-7-2,4,5-trimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl heptanoic acid, bronica tn, ccris 8939, +--7-3,5,6-trimethyl-1,4-benzoquinon-2-yl-7-phenylheptanoic acid, seratrodast jan/usan/inn
CC1=C(C(=O)C(=C(C1=O)C)C(CCCCCC(=O)O)C2=CC=CC=C2)C
354.446
354.44
10mg
C22H26O4
ZBVKEHDGYSLCCC-UHFFFAOYSA-N
7-phenyl-7-(2,4,5-trimethyl-3,6-dioxocyclohexa-1,4-dien-1-yl)heptanoic acid
2449
>98%
Videos