Learn More
AA 2414, Tocris Bioscience™
Thromboxane A2 (TP) receptor antagonist
POA
Description
Description
Thromboxane A2 (TP) receptor antagonist. Antiasthmatic agent; inhibits platelet aggregation and bronchoconstriction induced by a TXA2 mimetic in guinea pigs.