missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ KleenGuard™ A71 Chemical Spray Protection Coveralls
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Approved as Complex Design (Category 3) equipment - offering protection to the levels specified for Type 3, 4 and 5 by CEN
- Highly visible yellow colour - for improved worker safety
- Ideal for working with hazardous chemicals - protecting against liquid, aerosol and dry chemical splashes, sprays and dusts in low-pressure industrial cleaning and maintenance areas
- Single-use protective clothing to improve safety and comfort in high-risk areas - create a spray and liquid tight barrier against harmful chemical splashes, liquids or dust particles
- Suitable for use as protection against Infective Agents and Biohazards
- Suitable for use as protection against solid particles and particulate radioactive contamination
- Treated to be antistatic
- EN 1073-2 approved for radioactive dust protection
- Anti-static EN 1149-1 for critical areas
Specifications
Product Type | A71 Coveralls |
Size | 3X-Large |
Color | Yellow |
Cuff Style | Elastic |
Hood Type | Elastic |
For Use With (Application) | Hazardous Chemicals |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'