Learn More
Kimberly-Clark™ KLEENGUARD™ A45 Breathable Liquid and Particle Protection Hooded Coveralls
Single-use, breathable hooded coveralls with storm flap for respirator. The ultimate for user comfort, made from high-performance fabric and film laminate. Anti-static and general protection against water-based or chemical splashes, liquids or dust particles. PPE Category III.
Brand:  Kimberly-Clark™ 99680
Code : 94
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11700783
  • £260.00 / Pack of 25
Estimated Shipment
05-02-2018
Add to basket
Description
Description
  • Supplied in packs of 25
  • White, highly breathable, anti-static, silicone-free coveralls
  • High-performance fabric and film laminate construction for comfort, safety and strength.
  • Provides PPE Category III protection
  • Storm flap and hood designed for easier taping to a respirator
  • Provides anti-static and general protection against water-based or chemical splashes, liquids, and dust particles
  • The ultimate combination of protection and user comfort in more critical areas.
  • Hood specially designed to fit a respirator, longer arms,
  • Full-length zip
Specifications
Specifications
Two-way zip
25/PK
White
Specially Designed Hood
60cm
Breathable Liquid and Particle Protection Coverall
High-performance fabric and film laminate
Hooded
X-Large
Providing high levels of protection at the front and maintaining comfort and breathability at the back
10Kg
Videos
Documentation
Technical Documents