missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ARCTIC A28F Refrigerated Circulator

A28F refrigerated bath circulator provides efficient cooling for open and closed applications, with temperatures ranging from −28° to +200°C and 10L maximum bath volume.

£3122.74 - £4584.44

Specifications

Certifications/Compliance CE/ROHS/WEEE
Cooling Capacity 320w
Dimensions (L x W x H) 16.8 x 20.2 x 20.5 in. (42.6 x 51.2 x 52 cm)
Capacity (Metric) 10 L
Height (Metric) 52 cm
View More Specs

Products 7
Product Code Brand Flow Rate Controller Type Voltage Frequency Price Quantity & Availability  
Product Code Brand Flow Rate Controller Type Voltage Frequency Price Quantity & Availability  
10494759 View Documents Thermo Scientific™
152-4281
17L/min. SC100 230 V 50 Hz This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
11901875 View Documents Thermo Scientific™
1524286
17L/min. SC100 100 V 50/60 Hz This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
11620871 View Documents Thermo Scientific™
153-4281
17L/min. SC150 230 V 50 Hz This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
11640881 View Documents Thermo Scientific™
154-4281
17L/min. SC150L 230 V 50 Hz This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
11670901 View Documents Thermo Scientific™
155-4281
20L/min. AC150 230 V 50 Hz This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
11680921 View Documents Thermo Scientific™
156-4281
20L/min. AC200 230 V 50 Hz This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
12789715 View Documents Thermo Scientific™
157-4281
24L/min. PC200 230 V 50 Hz This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
Description

Description

Featuring superior cooling power, expansive temperature ranges, powerful force/suction pumps, and sophisticated digital control to ensure accuracy and reproducibility. Choose from three different controllers that allow you the flexibility to create the perfect fit for your application.
  • Energy-saving feature limits power consumption: tight control of cooling capacity dependent on application needs
  • Small refrigeration circulators are optimized for small footprint and low noise
  • Robust stainless-steel baths
  • Thermostat can be indexed 90° on all sides of the bath for easy viewing
  • External circulation set included for plumbing connection M16×1, 0.25 NPT male, 0.375in. O.D.
  • Three-side ventilation concept: up to two sides can be blocked with full refrigeration performance and no degradation of equipment
  • Draining port at the front
  • Integrated handles
  • CFC-free refrigerant

Includes:

Control cables, bridge with gasket and thumbscrews, fittings, clamps, pump plug for external circulation, 6ft. power cord, bath cover

Recommended for:

  • Calibration
  • Bioreactors
  • Rotary Evaporators
  • Condensers
  • Sample/Material Testing
  • Sample/Material Preparation
  • Warranty varies by country, please contact us for details
Specifications

Specifications

CE/ROHS/WEEE
16.8 x 20.2 x 20.5 in. (42.6 x 51.2 x 52 cm)
52 cm
35.6 kg
17.3 cm
320w
10 L
12.9 cm
51.2 cm