Learn More
Kimberly-Clark™ KLEENGUARD™ A25+ Hooded Splash Protection Coveralls
Provides general protection against water-based or chemical splashes, liquids or dust particles. Kimberly Clark™ KLEENGUARD™ Hooded A25+ Splash Protection Coveralls are perfect for general manufacturing and maintenance areas.
Brand:  Kimberly-Clark™ 89810
Code : NEW
Additional Details : Weight : 2.50000kg
 View more versions of this product
Product Code. 13418218
  • £149.00 / Pack of 25
Estimated Shipment
02-02-2018
Add to basket
Description
Description
  • Ideal for working in general manufacturing and maintenance
  • Offers complete freedom of movement with comfortable body design
  • Protects against powders, larger fibers, radioactive dust and chemical splashes
  • Silicone-free and anti-static
Specifications
Specifications
Full-length Zips
Elastic
Hooded
White
Elastic
Coveralls
128 to 136cm
Men's
Elastic
2X-Large
2.5Kg
Videos