missing translation for 'onlineSavingsMsg'
Learn More

A 841720, Tocris Bioscience™

Brand:  RD Systems (Bio-Techne) 3060/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13462324

  • £633.57 / 50mg


Estimated Shipment
28-02-2019
Add to basket

Specifications

Chemical Identifiers

869802-58-4
343.449
unii-u8c5c9d65s, 5-azepan-1-yl-13-dimethylamino-8-thia-3,5,10-triazatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2 7 ,3,9,11-pentaen-6-one, d06lyz, pyrido 3',2':4,5 thieno 3,2-d pyrimidin-4 3h-one, 9-dimethylamino-3-hexahydro-1h-azepin-1-yl, 3-azepan-1-yl-9-dimethylamino pyrido 1,2 thieno 3,4-d pyrimidin-4-one, 3-azepan-1-yl-9-dimethylamino pyrido 3',2':4,5 thieno 3,2-d pyrimidin-4 3h-one, 3-azepan-1-yl-9-dimethylamino-3h-pyrido 3,2:4,5 thieno 3,2-d pyrimidin-4-one, 9-dimethylamino-3-n-hexamethyleneiminyl-3h-5-thia-1,3,6-triazafluoren-4-one, 9-dimethylamino-3-hexahydro-1h-azepin-1-yl pyrido 3' ,2':4,5 thieno 3,2-d pyrimidin-4 3h-one, 9-dimethylamino-3-hexahydro-1h-azepin-1-yl pyrido 3',2':4,5 thieno 3,2-d pyrimidin-4 3h-one
3-(azepan-1-yl)-9-(dimethylamino)pyrido[1,2]thieno[3,4-d]pyrimidin-4-one
C17H21N5OS
GYWGXEGOXODOQU-UHFFFAOYSA-N
11559235
CN(C)C1=C2C3=C(C(=O)N(C=N3)N4CCCCCC4)SC2=NC=C1

Specifications

A 841720
869802-58-4
unii-u8c5c9d65s, 5-azepan-1-yl-13-dimethylamino-8-thia-3,5,10-triazatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2 7 ,3,9,11-pentaen-6-one, d06lyz, pyrido 3',2':4,5 thieno 3,2-d pyrimidin-4 3h-one, 9-dimethylamino-3-hexahydro-1h-azepin-1-yl, 3-azepan-1-yl-9-dimethylamino pyrido 1,2 thieno 3,4-d pyrimidin-4-one, 3-azepan-1-yl-9-dimethylamino pyrido 3',2':4,5 thieno 3,2-d pyrimidin-4 3h-one, 3-azepan-1-yl-9-dimethylamino-3h-pyrido 3,2:4,5 thieno 3,2-d pyrimidin-4-one, 9-dimethylamino-3-n-hexamethyleneiminyl-3h-5-thia-1,3,6-triazafluoren-4-one, 9-dimethylamino-3-hexahydro-1h-azepin-1-yl pyrido 3' ,2':4,5 thieno 3,2-d pyrimidin-4 3h-one, 9-dimethylamino-3-hexahydro-1h-azepin-1-yl pyrido 3',2':4,5 thieno 3,2-d pyrimidin-4 3h-one
CN(C)C1=C2C3=C(C(=O)N(C=N3)N4CCCCCC4)SC2=NC=C1
343.449
343.45
50mg
C17H21N5OS
GYWGXEGOXODOQU-UHFFFAOYSA-N
3-(azepan-1-yl)-9-(dimethylamino)pyrido[1,2]thieno[3,4-d]pyrimidin-4-one
11559235
>98%
Videos