missing translation for 'onlineSavingsMsg'
Learn More

A 804598, Tocris Bioscience™

Potent and selective P2X7 antagonist

Brand:  Tocris (Bio-Techne) 447350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13434814

  • £710.01 / 50mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, competitive P2X7 receptor antagonist (IC50 values are 8.9, 9.9 and 10.9 nM for mouse, rat and human P2X7 receptors respectively). Selective for P2X7 receptors (IC50 > 5-10 μM for a wide array of cell surface receptors and ion channels). Binds with high affinity (Ki app = 2.4 nM for rat P2X7 receptors).
Specifications

Chemical Identifiers

1125758-85-1
315.38
d0q1po, 1-cyano-2-1s-1-phenylethyl-3-quinolin-5-yl guanidine, n-cyano-n'-1s-1-phenylethyl-n'-5-quinolinyl-guanidine, n-cyano-n-1s-1-phenylethyl-n'-5-quinolinylguanidine, e-n'-cyano-n-1s-1-phenylethyl-n-quinolin-5-yl guanidine, n-cyano-n'-1s-1-phenylethyl-n-quinolin-5-yl guanidine
1-cyano-2-[(1S)-1-phenylethyl]-3-quinolin-5-ylguanidine
C19H17N5
PQYCRDPLPKGSME-AWEZNQCLSA-N
53325874
CC(C1=CC=CC=C1)N=C(NC#N)NC2=CC=CC3=C2C=CC=N3

Specifications

50mg
1125758-85-1
d0q1po, 1-cyano-2-1s-1-phenylethyl-3-quinolin-5-yl guanidine, n-cyano-n'-1s-1-phenylethyl-n'-5-quinolinyl-guanidine, n-cyano-n-1s-1-phenylethyl-n'-5-quinolinylguanidine, e-n'-cyano-n-1s-1-phenylethyl-n-quinolin-5-yl guanidine, n-cyano-n'-1s-1-phenylethyl-n-quinolin-5-yl guanidine
CC(C1=CC=CC=C1)N=C(NC#N)NC2=CC=CC3=C2C=CC=N3
315.38
315.37
A 804598
C19H17N5
PQYCRDPLPKGSME-AWEZNQCLSA-N
1-cyano-2-[(1S)-1-phenylethyl]-3-quinolin-5-ylguanidine
53325874
>99%
Videos
Documents